Learn More
Perkin Elmer US LLC Concave Tip for High Pressure Clamp
Catalog No. 502113537
Encompass_Preferred
Successful ATR analysis depends on how well the sample being analyzed comes into contact with the crystal. The High Pressure Clamp ensures the perfect amount of pressure. The clamp is shipped with three different tips: flat, concave, and swivel. Each tip is designed for specific types of sample.
Catalog No. 50-211-3537 Supplier Perkin Elmer US LLC Supplier No. L1272325
Successful ATR analysis depends on how well the sample being analyzed comes into contact with the crystal. The High Pressure Clamp ensures the perfect amount of pressure. The clamp is shipped with three different tips: flat, concave, and swivel. Each tip is designed for specific types of sample.