Learn More
DUBOIS CHEMICALS CONCENTRATE CLEAN SOL 12LT/CS
Catalog No. NC0162518
Encompass
The lab tech's best friend! Used on glassware, metals, plastics, ceramics, filter membranes and other hard surfaces. Cuts grease, oil foulants, and more at only 2% in water. Safe concentrated, mildly alkaline.
Catalog No. NC0162518 Supplier DUBOIS CHEMICALS Supplier No. M905012
The lab tech's best friend! Used on glassware, metals, plastics, ceramics, filter membranes and other hard surfaces. Cuts grease, oil foulants, and more at only 2% in water. Safe concentrated, mildly alkaline.