Learn More
Thermo Scientific CONCENTRATE FOR AN AS14
Catalog No. NC9752584 Shop All Thermo Scientific Products
Encompass
AS14 Eluent Concentrate (100x), for Anion and Cation Determinations, is a Ready-to-Dilute Eluent Concentrate for the IonPac AS14 Columns, Contains 0.35 M Carbonate/0.10 M Bicarbonate. It is Recommended for Fast, Reliable Eluent Preparation •Available as a 100X concentrate containing 0.80M carbonate/0.10M bicarbonate •Makes 25L of sodium carbonate/bicarbonate eluent •Prepared by the dissolution of high-purity chemicals in >18.2MOcm deionized water, which was tested by ion chromatography for ionic contaminants •Actual concentration is within ±2.5% and was determined by one or more proven reliable methods of analysis such as ion chromatography, UV spectrophotometry.
Catalog No. NC9752584 Supplier Thermo Scientific Supplier No. 053560
AS14 Eluent Concentrate (100x), for Anion and Cation Determinations, is a Ready-to-Dilute Eluent Concentrate for the IonPac AS14 Columns, Contains 0.35 M Carbonate/0.10 M Bicarbonate. It is Recommended for Fast, Reliable Eluent Preparation •Available as a 100X concentrate containing 0.80M carbonate/0.10M bicarbonate •Makes 25L of sodium carbonate/bicarbonate eluent •Prepared by the dissolution of high-purity chemicals in >18.2MOcm deionized water, which was tested by ion chromatography for ionic contaminants •Actual concentration is within ±2.5% and was determined by one or more proven reliable methods of analysis such as ion chromatography, UV spectrophotometry.