Learn More
MSC Rite-Kem Concentrated Cleaner, Industrial, pH 9.5-10
Catalog No. 502835616
Encompass
This concentrated cleaner is a clear, yellow liquid with a sassafrass odor, specifically formulated for industrial applications. It has a pH range of 9.5-10, making it effective for various cleaning tasks.
  • Clear, yellow liquid
  • Distinct sassafrass odor
  • pH of 9.5-10 for effective cleaning
  • Stable chemical composition
  • Will not undergo hazardous polymerization
  • Designed for industrial use only
Catalog No. 50-283-5616 Supplier MSC Supplier No. 75311753
This concentrated cleaner is a clear, yellow liquid with a sassafrass odor, specifically formulated for industrial applications. It has a pH range of 9.5-10, making it effective for various cleaning tasks.
  • Clear, yellow liquid
  • Distinct sassafrass odor
  • pH of 9.5-10 for effective cleaning
  • Stable chemical composition
  • Will not undergo hazardous polymerization
  • Designed for industrial use only