Learn More
Arkray Concentrated Washing Solution 3
SDP
Catalog No. 21403494
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
21-403-494 1 L
1 options
Warning: For in vitro diagnostic use only. Handle according to lab safety guidelines. Avoid contact with skin or eyes. Store as directed. Use protective equipment when preparing solutions.
Catalog No. 21-403-494 Supplier Arkray Supplier No. 101687
Add to Cart
Add to Cart
Warning: For in vitro diagnostic use only. Handle according to lab safety guidelines. Avoid contact with skin or eyes. Store as directed. Use protective equipment when preparing solutions.
This solution ensures thorough and consistent washing for A1c analyzers, reducing carryover and maintaining accuracy. Its concentrated formula saves storage space and simplifies preparation, providing reliable, efficient performance in every lab run.
  • Optimized Cleaning: Ensures thorough and consistent washing for A1c analyzers, minimizing sample carryover and cross-contamination.
  • High Accuracy: Maintains analyzer precision by effectively removing residual reagents, improving reliability of lab
Specifications
CE Marker CE-IVD
Clia Complexity High, Moderate
DoA Calibrators No
Type Solution
Disposable Single-use
Level 1
Quantity 1 L
Form Ready-to-Use (RTU)
Concentrated Washing Solution 3 delivers consistent and efficient cleaning for A1c analyzers, helping maintain accurate results and smooth lab workflow.