Learn More
Chondrex Inc Concentrating Solution
Catalog No. NC0904642
Encompass_Preferred
The concentration of collagen in culture media is generally low, therefore it is difficult to detect collagen in culture media samples within the standard range of this kit. Chondrex, Inc. recommends a sample concentration process using our Concentrating Solution (Cat # 90626).
Catalog No. NC0904642 Supplier Chondrex Inc Supplier No. 90626
The concentration of collagen in culture media is generally low, therefore it is difficult to detect collagen in culture media samples within the standard range of this kit. Chondrex, Inc. recommends a sample concentration process using our Concentrating Solution (Cat # 90626).