Learn More
BUCHI Concentrating Vessels Set for SyncorePlus Analyst
SDP
Catalog No. 05405917 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
SyncorePlus Analyst
Catalog No. For Use With (Equipment)
05-405-917 SyncorePlus Analyst
1 options
Catalog No. 05-405-917 Supplier BUCHI Supplier No. 046071
Add to Cart
Add to Cart
The chilled appendix of this analyst's tubes ensures maximum analyte recovery while concentrating it to a defined volume.
Safely and efficiently concentrate up to 12 samples to a defined residual volume.
  • 1 mL sample tubes
  • Protect heat-sensitive analytes with its cooled appendix
  • Achieve high analyte recoveries with the use of a flushback module
  • Rack Size: R-12
Specifications
For Use With (Application) Concentrating Samples To A Defined Volume
For Use With (Equipment) SyncorePlus Analyst
Quantity 12 Pk.
Volume (English) 0.03 oz.
Volume (Metric) 1 mL