Learn More
Chemglass Life Sciences Concentrator Tube, 10mL, Graduated, 19/22 Outer Joint, With Hooks, 0 to 1ml in 0.1 and 2 to 10ml in 1.0 Subdivisions
SDP
Catalog No. CG131602
Encompass_Preferred
Concentrator Tube, 10mL, Graduated, 19/22 Outer Joint, With Hooks, 0 to 1ml in 0.1 and 2 to 10ml in 1.0 Subdivisions. Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 100mm. Equipped with hooks.
Catalog No. CG131602 Supplier Chemglass Life Sciences Supplier No. CG131602
Concentrator Tube, 10mL, Graduated, 19/22 Outer Joint, With Hooks, 0 to 1ml in 0.1 and 2 to 10ml in 1.0 Subdivisions. Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 100mm. Equipped with hooks.