Learn More
Chemglass Life Sciences Concentrator Tube, 25mL, Graduated, 19/22 Outer Joint, With Out Hooks, 0 to 1ml in 0.1 and 2 to 25ml in 1.0 Subdivisions
SDP
Catalog No. 50997813
Encompass_Preferred
Concentrator Tube, 25mL, Graduated, 19/22 Outer Joint, With Out Hooks, 0 to 1ml in 0.1 and 2 to 25ml in 1.0 Subdivisions. Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 170mm. This item comes without hooks.
Catalog No. 50-997-813 Supplier Chemglass Life Sciences Supplier No. CG131612
Concentrator Tube, 25mL, Graduated, 19/22 Outer Joint, With Out Hooks, 0 to 1ml in 0.1 and 2 to 25ml in 1.0 Subdivisions. Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups. Tube can be stoppered for storage. Overall height is 170mm. This item comes without hooks.