Learn More
World Precision Instrument Concentric Bipolar Electrodes, Rhodes Style Tip-PTM321RC225
SDP
Catalog No. 502538622
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8622 Supplier World Precision Instrument Supplier No. PTM321RC225
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.