Learn More
World Precision Instrument Concentric Bipolar Electrodes with Extension, Rhodes Style Tip-PTM041RC225
SDP
Catalog No. 502538628
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8628 Supplier World Precision Instrument Supplier No. PTM041RC225
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.