Learn More
World Precision Instrument Concentric Bipolar Electrodes with Extension, Rhodes Style Tip-SSM041RC225
SDP
Catalog No. 502538565
Encompass
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.
Catalog No. 50-253-8565 Supplier World Precision Instrument Supplier No. SSM041RC225
The Concentric Bipolar Electrodes, designed for Neurological recording and stimulation and are ideal for acute applications. The Stainless Steel external tubing (E) provides support for accurate placement and manipulation during acute research.