Learn More
World Precision Instrument Concentric Bipolar Microelectrode, 76 mm, 0.013 inch
SDP
Catalog No. 502538621
Encompass
  • Platinum Iridium NS fine (3", 76mm)
  • 200K Impedence
  • 2-3μm tip
  • Package of 3
Catalog No. 50-253-8621 Supplier World Precision Instrument Supplier No. PTM3CC02INS
  • Platinum Iridium NS fine (3", 76mm)
  • 200K Impedence
  • 2-3μm tip
  • Package of 3