Learn More
Cayman Chemical ConcnavalIn A 250mg
SDP
Catalog No. 502969442
Encompass_Preferred
A plant lectin with diverse biological activities; induces erythrocyte agglutination, mitogenic stimulation, and has anticancer activity; has been used in affinity purification of various glycoproteins, as well as to induce T cell activation in vitro and liver injury in experimental mouse models of autoimmune hepatitis
Catalog No. 50-296-9442 Supplier Cayman Chemical Supplier No. 14951250 MG
A plant lectin with diverse biological activities; induces erythrocyte agglutination, mitogenic stimulation, and has anticancer activity; has been used in affinity purification of various glycoproteins, as well as to induce T cell activation in vitro and liver injury in experimental mouse models of autoimmune hepatitis