Learn More
Hach Company Conct Condct Alg Sens-Con 0.05
Catalog No. 5000207326 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant K 05- in NPT PVDF Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT PVDF Compression Fitting 15m (50ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07326 Supplier Hach Company Supplier No. 3422A1A50
Contacting-Conductivity Analog Sensor-Cell Constant K 05- in NPT PVDF Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT PVDF Compression Fitting 15m (50ft) Analog Cable Titanium Electrode