Learn More
Hach Company Conct Condct Alg Sens-Con 0.05
Catalog No. 5000207613 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT PVDF Compression FittingNominal Cell Constant (K) 0 05 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07613 Supplier Hach Company Supplier No. 3422A3C
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT PVDF Compression FittingNominal Cell Constant (K) 0 05 3/4 inch NPT PVDF Compression Fitting Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode