Learn More
Hach Company Conct Condct Alg Sens-Con 0.05
Catalog No. 5000207653 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT PVDF Compression FittingNominal Cell Constant k 0 05 cm-1 in NPT PVDF Compression Fitting 6 m (20 ft) Analogue Cable Titanium Electrode
Catalog No. 50-002-07653 Supplier Hach Company Supplier No. 3422A3A
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT PVDF Compression FittingNominal Cell Constant k 0 05 cm-1 in NPT PVDF Compression Fitting 6 m (20 ft) Analogue Cable Titanium Electrode