Learn More
Hach Company Conct Condct Alg Sens-Con 0.05
Catalog No. 5000207695 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT 316 Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT 316 Stainless Steel Compression Fitting 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07695 Supplier Hach Company Supplier No. 3422A2D
Contacting-Conductivity Analog Sensor-Cell Constant 0 05- in NPT 316 Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 1/2 inch NPT 316 Stainless Steel Compression Fitting 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode