Learn More
Hach Company Conct Condct Alg Sens-Con 0.05
Catalog No. 5000207700 Shop All Hach Company Products
Encompass
Contacting-Conductivity Analog Sensor-Cell Constant 0 05-3/4 inch NPT Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07700 Supplier Hach Company Supplier No. 3422A4A
Contacting-Conductivity Analog Sensor-Cell Constant 0 05-3/4 inch NPT Stainless Steel Compression FittingNominal Cell Constant (K) 0 05 3/4 inch NPT 316 Stainless Steel Compression Fitting 6m (20 ft) Analog Cable Titanium Electrode