Learn More
Ace Glass, Inc. Condenser, Allihn, 300mm, 24/40, #7 Ace-Thred connections, glass only
SDP
Catalog No. 50972658
Encompass_Preferred
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 24/40 standard taper inner and outer joints at the bottom and top, and a #7 Ace-Thred.
Catalog No. 50-972-658 Supplier Ace Glass, Inc. Supplier No. 594618
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 24/40 standard taper inner and outer joints at the bottom and top, and a #7 Ace-Thred.