Learn More
Ace Glass, Inc. Condenser, Allihn, 300mm, 29/42, #7 Ace-Thred connections glass only
SDP
Catalog No. 50972659
Encompass_Preferred
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 29/42 standard taper inner and outer joints at the bottom and top, and a #7 Ace-Thred.
Catalog No. 50-972-659 Supplier Ace Glass, Inc. Supplier No. 594622
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 29/42 standard taper inner and outer joints at the bottom and top, and a #7 Ace-Thred.