Learn More
Ace Glass, Inc. Condenser, Allihn, 24/25, micro Soxhlet
SDP
Catalog No. 50973417
Encompass_Preferred
Allihn condenser with 24/25 joint and approximately 139mm jacket length, for use with ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.
Catalog No. 50-973-417 Supplier Ace Glass, Inc. Supplier No. 677602
Allihn condenser with 24/25 joint and approximately 139mm jacket length, for use with ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.