Learn More
Cole-Parmer Hach 3422C3A Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=1.0
Catalog No. 50631139 Shop All Cole-Parmer Products
Encompass
Nonmetallic Ryton PPS cells 19065-55 and -57 have graphite electrode with 34" NPT(M) threads for mounting onto a 34" pipe tee or immersion applications. Sensor includes built-in 1000 Pt RTD and 20-ft (6-m) cable. PVDF cells 19065-60 through -68 have titanium electrodes with either 12" or 34" NPT(M) compression fitting for mounting onto a pipe tee with up to four inches of insertion depth. Reversing the compression fitting enables the sensor to be fastened onto the end of an immersion pipe. Cells include built-in 1000 Pt RTD and 20-ft (6-m) cable.
Catalog No. 50-631-139 Supplier Cole-Parmer Supplier No. 1906564
Nonmetallic Ryton PPS cells 19065-55 and -57 have graphite electrode with 34" NPT(M) threads for mounting onto a 34" pipe tee or immersion applications. Sensor includes built-in 1000 Pt RTD and 20-ft (6-m) cable. PVDF cells 19065-60 through -68 have titanium electrodes with either 12" or 34" NPT(M) compression fitting for mounting onto a pipe tee with up to four inches of insertion depth. Reversing the compression fitting enables the sensor to be fastened onto the end of an immersion pipe. Cells include built-in 1000 Pt RTD and 20-ft (6-m) cable.