Learn More
Azzota Corp Cond Electrode K 0.1 Pt glass
Catalog No. 5000887269
Encompass_Preferred
CON-0 1 Conductivity Electrode Platinum conductivity electrode suitable for general applications range 0 to 100 S/cm Platinum cell conductivity electrode For measuring the pure water or low conductivity liquids Cell constant (K) 0 1 6 holes connector
Catalog No. 50-008-87269 Supplier Azzota Corp Supplier No. CON0.1
CON-0 1 Conductivity Electrode Platinum conductivity electrode suitable for general applications range 0 to 100 S/cm Platinum cell conductivity electrode For measuring the pure water or low conductivity liquids Cell constant (K) 0 1 6 holes connector