Learn More
Cole-Parmer Jenway 4510 Conductivity Meter with Probe Stand; 120 V
Supplier:  Cole-Parmer 3544002
Encompass_Preferred
Get accurate readings quickly with this meter. Always measure at the best resolution with automatic standard recognition. Easily calibrate with either standard solution or known cell constant.
Catalog No. 50-196-1247