Learn More
Hach Company Conductivity Standard Solution, 12.88 mS/cm, KCl, 125mL
Supplier:  Hach Company LZW9721.99
Encompass
Conductivity Standard Solution. 12.88 mS/cm ±0.11 mS/cm, as Potassium Chloride (KCl) (0.1M). 125 mL bottle.
Catalog No. 50-199-8036