Learn More
Hach Company Conductivity Standard Solution, 12.88 mS/cm, KCl, 250mL
Catalog No. 501998035 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. 12.88 mS/cm ±0.11 mS/cm at 25°C, as Potassium Chloride (KCl) (0.1M). 250 mL bottle.
Catalog No. 50-199-8035 Supplier Hach Company Supplier No. LZW9720.99
Conductivity Standard Solution. 12.88 mS/cm ±0.11 mS/cm at 25°C, as Potassium Chloride (KCl) (0.1M). 250 mL bottle.