Learn More
Hach Company Conductivity Standard Solution, 1413 µS/cm, KCl, 250mL
Catalog No. 501998033 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. 1413 µS/cm ±12 µS/cm at 25°C, as Potassium Chloride (KCl) (0.01M). 250 mL bottle.
Catalog No. 50-199-8033 Supplier Hach Company Supplier No. LZW9710.99
Conductivity Standard Solution. 1413 µS/cm ±12 µS/cm at 25°C, as Potassium Chloride (KCl) (0.01M). 250 mL bottle.