Learn More
Hach Company Conductivity Standard Solution, 1413 µS/cm, KCl, 125mL
Supplier:  Hach Company LZW9711.99
Encompass
Conductivity Standard Solution. 1413 µS/cm ±12 µS/cm at 25°C, as Potassium Chloride (KCl) (0.01M). 125 mL bottle.
Catalog No. 50-199-8034