Learn More
Hach Company Conductivity Standard Solution, 147 µS/cm, KCl, 250mL
Supplier:  Hach Company LZW9700.99
Encompass
Conductivity Standard Solution. 147 µS/cm ±5 µS/cm at 25°C, as Potassium Chloride (KCl) (0.001M). 250 mL bottle.
Catalog No. 50-199-8031