Learn More
Hach Company Condct. Stand Sol 12.88mS/cm
Catalog No. 5000207011 Shop All Hach Company Products
Encompass
Conductivity Standard Solution 12 88 mS/cm 1% at 25 C as Potassium Chloride (KCl) (0 1M) 500 mL bottle
Catalog No. 50-002-07011 Supplier Hach Company Supplier No. C20C250
Conductivity Standard Solution 12 88 mS/cm 1% at 25 C as Potassium Chloride (KCl) (0 1M) 500 mL bottle