Learn More
Caron Products And Services CONDENSATE RECIRCULATING SYS
Catalog No. NC2928754
Encompass_Preferred
  • Recycles condensate and utilizes tap water, reducing water consumption.
  • Delivers lab-quality water, preventing mineral deposits.
  • Employs a multi-stage filtration system with an Ionic Silver Stick to eliminate bacteria and viruses.
  • Features single-switch operation and automatic air purge for easy maintenance.
  • Allows for placement in labs without dedicated floor drains.
  • Equipped with front/rear handles and casters for easy relocation.
  • Visual indicators for low water level and filter replacement.
  • Full-length side access panels for convenient servicing.
  • Water reservoir capacity of 7 gal (26.5 L).
  • Electrical specifications of 115V, 60 Hz, 5A.
  • Exterior dimensions of 14 in x 26.6 in x 26.5 in (35.5 x 67.6 x 67.3 cm).
  • Weighs 84 lbs (38 kg) when empty.
  • Compatible with CARON humidity controlled chambers.
Catalog No. NC2928754 Supplier Caron Products And Services Supplier No. CRSY1021
Add to Cart
Add to Cart
Request a Quote
  • Recycles condensate and utilizes tap water, reducing water consumption.
  • Delivers lab-quality water, preventing mineral deposits.
  • Employs a multi-stage filtration system with an Ionic Silver Stick to eliminate bacteria and viruses.
  • Features single-switch operation and automatic air purge for easy maintenance.
  • Allows for placement in labs without dedicated floor drains.
  • Equipped with front/rear handles and casters for easy relocation.
  • Visual indicators for low water level and filter replacement.
  • Full-length side access panels for convenient servicing.
  • Water reservoir capacity of 7 gal (26.5 L).
  • Electrical specifications of 115V, 60 Hz, 5A.
  • Exterior dimensions of 14 in x 26.6 in x 26.5 in (35.5 x 67.6 x 67.3 cm).
  • Weighs 84 lbs (38 kg) when empty.
  • Compatible with CARON humidity controlled chambers.