Learn More
Ace Glass, Inc. Condenser, Allihn, 200mm, 24/25 st joints top & bottom
SDP
Catalog No. 50972656
Encompass_Preferred
200mm Allihn condenser with approximately one bulb per 50mm. Features 24/25 standard taper joints at top and bottom.
Catalog No. 50-972-656 Supplier Ace Glass, Inc. Supplier No. 594534
200mm Allihn condenser with approximately one bulb per 50mm. Features 24/25 standard taper joints at top and bottom.