Learn More
Chemglass Life Sciences Condenser, Reflux, 415mm, 24/40 Joint, 300mm Jacket Length
SDP
Catalog No. CG121605
Encompass_Preferred
Condenser, Reflux, 415mm, 24/40 Joint, 300mm Jacket Length. Reflux condenser with the coolant circulated through the inner coil instead of through the jacket as in the Graham style condenser. Condenser has standard taper top outer and lower inner drip tip joints. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121605 Supplier Chemglass Life Sciences Supplier No. CG121605
Condenser, Reflux, 415mm, 24/40 Joint, 300mm Jacket Length. Reflux condenser with the coolant circulated through the inner coil instead of through the jacket as in the Graham style condenser. Condenser has standard taper top outer and lower inner drip tip joints. Hose connections have an O.D. of 10mm at the middle serration.