Learn More
Hach Company Condenser
Supplier:  Hach Company 2265000
Encompass
Designed for use with the General Purpose Distillation Apparatus. Round bottom, 500 mL capacity. Long neck with branch and thermometer port. Neck extends 90 mm above flask body and branch extends 150 mm above flask body. Both neck and branch have 20-400 screw-thread finish. Thermometer port has 13-425 screw-thread finish.
Catalog No. 50-199-5671