Learn More
Ace Glass, Inc. Condenser, 250mm, 24/40 st joints top and bottom, #7 Ace-Thred connections, complete with Ace-Safe connectors
SDP
Catalog No. 50972673
Encompass_Preferred
250mm double-jacketed, coiled condenser with standard 24/40 inner and outer joint at bottom and top. Includes #7 Ace-Thred and Ace-Safe PTFE hose connections on inlet and outlet with barb for I.D. tubing. Provides extra cooling capacity for volatile materials.
Catalog No. 50-972-673 Supplier Ace Glass, Inc. Supplier No. 5956143
250mm double-jacketed, coiled condenser with standard 24/40 inner and outer joint at bottom and top. Includes #7 Ace-Thred and Ace-Safe PTFE hose connections on inlet and outlet with barb for I.D. tubing. Provides extra cooling capacity for volatile materials.