Learn More
Chemglass Life Sciences Condenser, Reflux, 390mm, 24/40 Joint, 250mm Jacket Length
SDP
Catalog No. CG1215A10
Encompass_Preferred
Condenser, Reflux, 390mm, 24/40 Joint, 250mm Jacket Length. Reflux condenser having an internal coil that is water jacketed. Vapors are condensed both on the coil and the inner wall of the jacket. The top end has a standard taper outer joint and the lower end has an inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG1215A10 Supplier Chemglass Life Sciences Supplier No. CG1215A10
Condenser, Reflux, 390mm, 24/40 Joint, 250mm Jacket Length. Reflux condenser having an internal coil that is water jacketed. Vapors are condensed both on the coil and the inner wall of the jacket. The top end has a standard taper outer joint and the lower end has an inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.