Learn More
Ace Glass, Inc. Condenser, 255mm, extraction, Allihn, 34/45
SDP
Catalog No. 50978877
Encompass_Preferred
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 34/45 joint. Jacket length is approximately 255mm with one bulb per 50mm.
Catalog No. 50-978-877 Supplier Ace Glass, Inc. Supplier No. 674002
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 34/45 joint. Jacket length is approximately 255mm with one bulb per 50mm.