Learn More
Ace Glass, Inc. Condenser, 255mm, extraction, Allihn, 45/50
SDP
Catalog No. 50973414
Encompass_Preferred
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 45/50 joint. Jacket length is approximately 255mm with one bulb per 50mm.
Catalog No. 50-973-414 Supplier Ace Glass, Inc. Supplier No. 674006
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 45/50 joint. Jacket length is approximately 255mm with one bulb per 50mm.