Learn More
Ace Glass, Inc. Condenser, Allihn, 300mm, 24/40 st joints top and bottom, #7 Ace-Safe connectors included
SDP
Catalog No. 50972660
Encompass_Preferred
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 24/40 standard taper inner and outer joints at the bottom and top, a #7 Ace-Thred, and Ace-Safe PTFE hose connections on inlet and outlet with barb for I.D. tubing.
Catalog No. 50-972-660 Supplier Ace Glass, Inc. Supplier No. 5946118
300mm Allihn, bulb type, condenser with approximately one bulb per 50mm. Condenser features 24/40 standard taper inner and outer joints at the bottom and top, a #7 Ace-Thred, and Ace-Safe PTFE hose connections on inlet and outlet with barb for I.D. tubing.