Learn More
Chemglass Life Sciences Coil Condenser, "A" Assembly, Rotary Evaporator, Plastic Coated
SDP
Catalog No. CG132610
Encompass_Preferred
Coil Condenser, "A" Assembly, Rotary Evaporator, Plastic Coated. Double coil condenser for use with rotary evaporators for cooling with tap water in a diagonal position. Condenser has a 50mm O.D. flange at the bottom, a 19/38 top valve, and removable hose connections using GL-14 glass threads. Condenser is supplied complete with plastic caps and serrated hose connections.
Catalog No. CG132610 Supplier Chemglass Life Sciences Supplier No. CG132610
Coil Condenser, "A" Assembly, Rotary Evaporator, Plastic Coated. Double coil condenser for use with rotary evaporators for cooling with tap water in a diagonal position. Condenser has a 50mm O.D. flange at the bottom, a 19/38 top valve, and removable hose connections using GL-14 glass threads. Condenser is supplied complete with plastic caps and serrated hose connections.