Learn More
Chemglass Life Sciences Condenser, Graham, 380mm, 24/40 Joint, 250mm Jacket Length
SDP
Catalog No. CG121403
Encompass_Preferred
Condenser, Graham, 380mm, 24/40 Joint, 250mm Jacket Length. Condenser has a top standard taper outer and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121403 Supplier Chemglass Life Sciences Supplier No. CG121403
Condenser, Graham, 380mm, 24/40 Joint, 250mm Jacket Length. Condenser has a top standard taper outer and lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.