Learn More
Ace Glass, Inc. Condenser, 390mm, extraction, Allihn, 45/50, for 6846 extraction apparatus
SDP
Catalog No. 50973416
Encompass_Preferred
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 45/50 joint. Jacket length is approximately 390mm.
Catalog No. 50-973-416 Supplier Ace Glass, Inc. Supplier No. 674026
Allihn bulb-type extraction condenser for use with regular extraction apparatus. Permits greater condensing capacity. Condenser has a 45/50 joint. Jacket length is approximately 390mm.