Learn More
Ace Glass, Inc. Condenser, Allihn, 600mm, 24/40 st joints top & bottom
SDP
Catalog No. 50972652
Encompass_Preferred
600mm Allihn condenser with approximately one bulb per 50mm. Features 24/40 standard taper joints at top and bottom.
Catalog No. 50-972-652 Supplier Ace Glass, Inc. Supplier No. 594517
600mm Allihn condenser with approximately one bulb per 50mm. Features 24/40 standard taper joints at top and bottom.