Learn More
Ace Glass, Inc. Condenser, Allihn, 600mm, 29/42 st joints top & bottom
SDP
Catalog No. 50972655
Encompass_Preferred
600mm Allihn condenser with approximately one bulb per 50mm. Features 29/42 standard taper joints at top and bottom.
Catalog No. 50-972-655 Supplier Ace Glass, Inc. Supplier No. 594527
600mm Allihn condenser with approximately one bulb per 50mm. Features 29/42 standard taper joints at top and bottom.