Learn More
Wilmad Labglass CONDENSER AIR REFLUX 1410
Supplier:  Wilmad Labglass ML4000600
Encompass_Preferred
Condenser, Air Reflux. Effective Length: 100mm; Lower Joint Size: [ST] 14/10 Inner; Upper Joint Size: [ST] 14/10 Inner. Condenser uses surrounding air to condense vapors during refluxing
Catalog No. 50-802-888