Learn More
Chemglass Life Sciences Condenser, Air Reflux, 7/10-14/10 Joint Size, Minum-Ware
SDP
Catalog No. 501532692
Encompass_Preferred
Condenser, Air Reflux, 7/10-14/10 Joint Size, Minum-Ware.
Catalog No. 50-153-2692 Supplier Chemglass Life Sciences Supplier No. MW6402
Condenser, Air Reflux, 7/10-14/10 Joint Size, Minum-Ware.