Learn More
Chemglass Life Sciences Condenser, Allihn, 430mm, 24/40 Joint, 300mm Jacket Length, Removable Hose Connection
SDP
Catalog No. CG1206HC05
Encompass_Preferred
Condenser, Allihn, 430mm, 24/40 Joint, 300mm Jacket Length, Removable Hose Connection. Allihn style reflux condenser similar to CG-1206 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.
Catalog No. CG1206HC05 Supplier Chemglass Life Sciences Supplier No. CG1206HC05
Condenser, Allihn, 430mm, 24/40 Joint, 300mm Jacket Length, Removable Hose Connection. Allihn style reflux condenser similar to CG-1206 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.