Learn More
Chemglass Life Sciences Condenser, Allihn, 380mm, 24/40 Joint, 250mm Jacket Length
SDP
Catalog No. CG120603
Encompass_Preferred
Condenser, Allihn, 380mm, 24/40 Joint, 250mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG120603 Supplier Chemglass Life Sciences Supplier No. CG120603
Condenser, Allihn, 380mm, 24/40 Joint, 250mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.