Learn More
Chemglass Life Sciences Condenser, Allihn, 445mm, 24/40 Inner Joint, 300mm Jacket Length
SDP
Catalog No. CG120703
Encompass_Preferred
Condenser, Allihn, 445mm, 24/40 Inner Joint, 300mm Jacket Length. Allihn style reflux condenser with a plain tubulation at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.• NOTE: See CG-1368 for Allihn Condensers used with soxhlet extraction apparatus.
Catalog No. CG120703 Supplier Chemglass Life Sciences Supplier No. CG120703
Condenser, Allihn, 445mm, 24/40 Inner Joint, 300mm Jacket Length. Allihn style reflux condenser with a plain tubulation at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.• NOTE: See CG-1368 for Allihn Condensers used with soxhlet extraction apparatus.